missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SERPINA9 Monoclonal Antibody (585302)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA561552
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
SERPINA9 is a protease inhibitor that inhibits trypsin and trypsin-like serine proteases (in vitro). This protein inhibits plasmin and thrombin with lower efficiency (in vitro).
Specifications
| SERPINA9 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| Q86WD7 | |
| SERPINA9 | |
| Mouse myeloma cell line NS0-derived recombinant human Serpin A9/Centerin, Ala24-Thr417. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Immunohistochemistry (Paraffin) | |
| 585302 | |
| Unconjugated | |
| SERPINA9 | |
| Centerin; GCET1; Germinal center B-cell-expressed transcript 1 protein; serine (or cysteine) proteinase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 9; serine proteinase inhibitor A11; serpin A9; serpin family A member 9; serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 9; SERPINA11; SERPINA11b; SERPINA9; UNQ692/PRO1337 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 327657 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction