missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Serine Palmitoyltransferase 2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98540-50ul
This item is not returnable.
View return policy
Description
Serine Palmitoyltransferase 2 Polyclonal antibody specifically detects Serine Palmitoyltransferase 2 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Serine Palmitoyltransferase 2 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin 1:50-1:200 | |
| EC 2.3.1, EC 2.3.1.50, KIAA0526HSN1C, LCB 2, LCB2a, LCB2NSAN1C, Long chain base biosynthesis protein 2, Long chain base biosynthesis protein 2a, serine palmitoyltransferase 2, serine palmitoyltransferase, long chain base subunit 2, serine palmitoyltransferase, subunit II, Serine-palmitoyl-CoA transferase 2, SPT 2, SPT2 | |
| Produced in rabbits immunized with E. coli-derived Human Serine Palmitoyltransferase 2 fragment. | |
| 50 μg | |
| Cancer, Cardiovascular Biology, Endocrinology, Signal Transduction | |
| 9517 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.0) | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction