missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
The antibody is specific to SERINC1.
SERINC1, also named as KIAA1253, TDE1L and TDE2, belongs to the TDE1 family. SERINC1 enhances the incorporation of serine into phosphatidylserine and sphingolipids.
Specifications
Specifications
| Antigen | SERINC1 |
| Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.39 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | SERINC1 |
| Gene Accession No. | Q9NRX5 |
| Gene Alias | KIAA1253, SERINC1, serine incorporator 1, TDE1L, TDE2, TMS 2 |
| Gene Symbols | SERINC1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?