Learn More
Thermo Scientific™ Septa
Product Code. 15360441 Shop All Thermo Scientific Products
missing translation for 'orderingAttributeHoverText'
Description:
BTO
Marathon
TR-Blue
TR-Green
Diameter (Metric) Inner:
9 mm
11 mm
11.5 mm
12.7 mm
17 mm
Temperature (Metric) Max:
200°C to 250°C
300°C
350°C
400°C
Quantity:
48 (Blister Pk.)
50 (Blister Pk.)
50 (Glass Jar)
missing translation for 'unitSize'
Each
Pack of 48
Pack of 50
Product Code. 15360441
missing translation for 'mfr': Thermo Scientific™ 313P3215BP
Only null left
Add to basket
Add to basket
Use low bleed, robust BTO Septa for demanding GC-MS applications.
Ensure optimal performance of your GC instrument with bleed and temperature optimized Thermo Scientific™ BTO and TR-Green Septa. Made of low-bleed silicone, these septa have excellent mechanical properties, are ideal for demanding GC-MS applications, and may be used reliably up to 400 °C.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
TRUSTED_SUSTAINABILITY
Specifications
Type Low Coring Septum
Description Marathon
Temperature (Metric) Max 300°C
For Use With (Equipment) Trace and Focus GC Instruments
Material Silicone
Diameter (Metric) Inner 17 mm
Quantity 48 (Blister Pk.)