Learn More
Thermo Scientific™ Septa
Product Code. 15294543 Shop All Thermo Scientific Products
Click to view available options
Description:
BTO
Marathon
TR-Blue
TR-Green
Diameter (Metric) Inner:
9 mm
11 mm
11.5 mm
12.7 mm
17 mm
Temperature (Metric) Max:
200°C to 250°C
300°C
350°C
400°C
Quantity:
48 (Blister Pk.)
50 (Blister Pk.)
50 (Glass Jar)
Unit Size:
Each
Pack of 48
Pack of 50
Product Code. 15294543
Brand: Thermo Scientific™ 313B3211
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Use low bleed, robust BTO Septa for demanding GC-MS applications.
Ensure optimal performance of your GC instrument with bleed and temperature optimized Thermo Scientific™ BTO and TR-Green Septa. Made of low-bleed silicone, these septa have excellent mechanical properties, are ideal for demanding GC-MS applications, and may be used reliably up to 400 °C.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
TRUSTED_SUSTAINABILITY
Specifications
Type Low Coring Silicone
Description TR-Blue
Temperature (Metric) Max 200°C to 250°C
For Use With (Equipment) Trace and Focus GC Instruments
Material Silicone
Diameter (Metric) Inner 17 mm
Quantity 50 (Glass Jar)