missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Senataxin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 6 publications
Brand: Novus Biologicals NB100-57542
This item is not returnable.
View return policy
Description
Senataxin Polyclonal specifically detects Senataxin in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Chromatin Immunoprecipitation (ChIP), Knockdown Validated.
Specifications
| Senataxin | |
| Polyclonal | |
| Unconjugated | |
| Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
| ALS4recessive, non-Friedreich type 1, AOA2FLJ43459, DKFZp781B151, KIAA0625FLJ12840, senataxin, SETX | |
| Rabbit | |
| 303 kDa | |
| 100 μL | |
| Primary | |
| Based on 100% sequence identity, this antiboday is predicted to react with Gorilla and Norhern white-cheeked gibbon. | |
| Store at 4C. Do not freeze. |
| ChIP Assay, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, KnockDown | |
| 1.0 mg/mL | |
| Western Blot 1:2000-1:10000, Immunocytochemistry/Immunofluorescence, Immunoprecipitation 2 - 10 μg/mg lysate, Chromatin Immunoprecipitation (ChIP) | |
| Q7Z333 | |
| SETX | |
| The immunogen recognized by this antibody maps to a region between residue 875 and 925 of human senataxin using the numbering given in entry NP_055861.2 (GeneID 23064). | |
| Affinity Purified | |
| RUO | |
| 23064 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction