missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SEMA6C Monoclonal Antibody (256631)

Rat Monoclonal Antibody
Brand: Invitrogen™ MA561101
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
This gene product is a member of the semaphorin family of proteins. Semaphorins represent important molecular signals controlling multiple aspects of the cellular response that follows CNS injury, and thus may play an important role in neural regeneration.
Specifications
| SEMA6C | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| Q9WTM3 | |
| SEMA6C | |
| Mouse myeloma cell line NS0-derived recombinant mouse Semaphorin 6C, Ala26-Ile602. | |
| 500 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG2a |
| Immunohistochemistry (Paraffin) | |
| 256631 | |
| Unconjugated | |
| SEMA6C | |
| KIAA1869; mKIAA1869; m-Sema Y; m-SemaY; m-SemaY2; sema domain, transmembrane domain (TM), and cytoplasmic domain, (semaphorin) 6C; sema Y; SEMA6C; semaphorin 6c; semaphorin-6C; Semaphorin-Y; Semay | |
| Rat | |
| Protein A/G | |
| RUO | |
| 20360 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction