missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SEMA4A Polyclonal Antibody
Sheep Polyclonal Antibody
Brand: Invitrogen™ PA5148887
This item is not returnable.
View return policy
Description
Reconstitute with sterile PBS to a final concentration of 0.2 mg/mL.
SEMA4A is a member of the semaphorin family of soluble and transmembrane proteins. Semaphorins are involved in guidance of axonal migration during neuronal development and in immune responses.
Specifications
| SEMA4A | |
| Polyclonal | |
| Unconjugated | |
| SEMA4A | |
| AI132332; CORD10; RP35; Sema B; sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and short cytoplasmic domain, (semaphorin) 4A; Sema4a; Semab; semaphorin 4A; semaphorin B; semaphorin-4A; Semaphorin-B; SemB; UNQ783/PRO1317 | |
| Sheep | |
| Antigen affinity chromatography | |
| RUO | |
| 64218 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Western Blot | |
| 0.2 mg/mL | |
| PBS with trehalose and no preservative | |
| Q9H3S1 | |
| SEMA4A | |
| Mouse myeloma cell line NS0-derived recombinant human Semaphorin 4A, Gly32-His683. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction