missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SEMA3E Monoclonal Antibody (400520)
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA561281

Product Code. 30535917

  • 459.00€ / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Reconstitute at 0.5 mg/mL in sterile PBS.

The Semaphorins make up the largest family of axon guidance cues yet described. Semaphorins are divided into 8 classes (classes 3-7 found in vertebrates). Class 3 Semaphorins are secreted, classes 4 through 6 are transmembrane proteins, and class 7 are membrane associated via glycosylphosphatidylinositol (GPI) linkage. They are characterized structurally by a conserved approximately 400 amino acid sema domain. They are classically described as collapsing factors and mediators of axon repulsion, although they may also act as context-dependent chemoattractants. Semaphorins have been shown to have roles in cardiovascular development and in the regulation of immune cell antigen presentation. Receptors or receptor complexes that mediate semaphorin signaling include proteins of the Neuropilin and Plexin families.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SEMA3E
Monoclonal
0.5 mg/mL
PBS with trehalose and no preservative
O15041
SEMA3E
Mouse myeloma cell line NS0-derived recombinant human Semaphorin 3E, Thr25-Ser775 (Arg557Ala and Arg560Ala).
100 μg
Primary
Human
Antibody
IgG1
Western Blot
400520
Unconjugated
SEMA3E
6430702L12; AA408817; coll-5; KIAA0331; mKIAA0331; M-sema H; M-SEMAH; M-SemaK; sema domain, immunoglobulin domain (Ig), short basic domain, secreted, (semaphorin) 3E; sema H; Sema3e; SEMAH; semaphorin 3E; semaphorin-3E; Semaphorin-H; Semh
Mouse
Protein A/G
RUO
9723
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.