missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Selenium for AAS 10g/L HNO3 matrix, Fisher Chemical™
Shop All Fisher Chemical Products

Click to view available options
Quantity:
100 mL
Unit Size:
100mL
Specifications
Specifications
| Packaging | Bottle |
| CAS | 7697-37-2 |
| Physical Form | Liquid |
| Molecular Formula | HNO3 |
| Concentration | 10 g/L ±0.02 g/L |
| MDL Number | MFCD00011349 |
| UN Number | 1760 |
| InChI Key | GRYLNZFGIOXLOG-UHFFFAOYSA-N |
| Type | Single-element Concentrated Standard Solution |
| SMILES | O[N+]([O-])=O |
| Show More |
Safety and Handling
Product Identifier
- Single-element Standard Solution for AAS. Selenium (Se) 10000mg/l in HNO3 5%
- Danger
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity after repeated exposure Category 2
- LONG-TERM AQUATIC HAZARD Chronic 3
- H314-Causes severe skin burns and eye damage.
- H373-May cause damage to organs.
- H412-Harmful to aquatic life with long lasting effects.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P314-Get medical advice/attention if you feel unwell.
- P501-Dispose of contents/container to .
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction