Learn More
SEC24D Antibody (S01-5G5), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-19915-50ul
This item is not returnable.
View return policy
Description
SEC24D Monoclonal antibody specifically detects SEC24D in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| SEC24D | |
| Monoclonal | |
| 0.3 mg/mL | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 9871 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| S01-5G5 | |
| Unconjugated | |
| FLJ43974, KIAA0755member D, SEC24 family, member D (S. cerevisiae), SEC24 related gene family, member D | |
| A synthetic peptide of human SEC24D (Uniprot # O94855) | |
| 50 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |