missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Seasonal H1N1 Hemagglutinin Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
168.00€ - 507.00€
Specifications
| Antigen | Seasonal H1N1 Hemagglutinin |
|---|---|
| Concentration | 1 mg/mL |
| Applications | ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18616555
|
Novus Biologicals
NBP2-41106-0.025mg |
0.025 mg |
168.00€
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18164378
|
Novus Biologicals
NBP2-41106 |
0.1 mg |
507.00€
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Seasonal H1N1 Hemagglutinin Polyclonal specifically detects Seasonal H1N1 Hemagglutinin in Influenza A Virus H1N1, Virus samples. It is validated for ELISA.Specifications
| Seasonal H1N1 Hemagglutinin | |
| ELISA | |
| Unconjugated | |
| RUO | |
| PBS with 0.02% Sodium Azide | |
| Antibody was raised against a synthetic peptide from the Hemagglutinin protein. The peptide sequence is unique from the peptide sequence for product NBP2-41104 and NBP2-41108. This antibody is a cognate pair with antibody NBP2-41107. The immunogen is located within amino acids 180 - 230 of Seasonal H1N1 Hemagglutinin. Amino Acid Squence: PNIGDQKALTHTEN | |
| Primary | |
| This antibody is specific for the seasonal H1N1 influenza Hemagglutinin and will not recognize the corresponding Hemagglutinin sequence from the swine-origin H1N1 influenza (A/California/14/2009 (H1N1)). Will not cross-react with peptide corresponding to the swine-origin H1N1 influenza Hemagglutinin. |
| 1 mg/mL | |
| Polyclonal | |
| Rabbit | |
| Influenza virus, Virus | |
| Hemagglutinin, Hemagglutinin HA1 chain, Hemagglutinin HA2 chain | |
| IgG | |
| Affinity Purified | |
| 68 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title