missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SDHB Rabbit anti-Human, Clone: SDHB/6697R, Novus Biologicals™
Rabbit Monoclonal Antibody
528.00€
Specifications
| Antigen | SDHB |
|---|---|
| Clone | SDHB/6697R |
| Concentration | 0.2 mg/mL |
| Dilution | Immunohistochemistry-Paraffin 1 to 2 μg/mL |
| Applications | Immunohistochemistry (Paraffin) |
Description
SDHB Monoclonal specifically detects SDHB in Human samples. It is validated for Immunohistochemistry-Paraffin.Specifications
| SDHB | |
| 0.2 mg/mL | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cancer | |
| 10 mM PBS with 0.05% BSA | |
| 6390 | |
| IgG κ | |
| Protein A or G purified |
| SDHB/6697R | |
| Immunohistochemistry-Paraffin 1 to 2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| EC 1.3.5.1, FLJ92337, IP, Iron-sulfur subunit of complex II, PGL4, SDH, SDH1, SDH2, SDHIP, succinate dehydrogenase [ubiquinone] iron-sulfur subunit, mitochondrial, succinate dehydrogenase complex, subunit B, iron sulfur (Ip) | |
| Synthetic peptide corresponding to residues in human SDHB was used as an immunogen. | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title