missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ SDHA Antibody (SDHA/7491), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-26729-100ug
This item is not returnable.
View return policy
Description
SDHA Monoclonal antibody specifically detects SDHA in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| SDHA | |
| Monoclonal | |
| 0.2 mg/mL | |
| Immunohistochemistry-Paraffin 1-2 μg/mL for 30 minutes at RT | |
| CMD1GG, flavoprotein subunit of complex II, FP, MC2DN1, NDAXOA, PGL5, PPGL5, SDH1, SDH2, SDHF, succinate dehydrogenase complex, subunit A, flavoprotein (Fp) | |
| Recombinant fragment (around aa 450-650) of human SDHA protein (exact sequence is proprietary) | |
| 100 μg | |
| Hypoxia | |
| 6389 | |
| Store at 4°C. | |
| IgG2 κ |
| Immunohistochemistry (Paraffin) | |
| SDHA/7491 | |
| Unconjugated | |
| 10 mM PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction