Learn More
SDC2 Mouse anti-Human, Unconjugated, Clone: 3A7B2, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67088-1-IG-20UL
 View more versions of this product
Product Code. 16829515
  • 145.00€ / 20µL
Description
The SDC2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human SDC2. This antibody recognizes human antigen. The SDC2 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
SDC2
Monoclonal
1000 μg/mL
Mouse
Primary
Human
Western Blot, Immunohistochemistry, ELISA
3A7B2
Unconjugated
20 μL
6383
IgG2a