Learn More
SDC2 Mouse anti-Human, Unconjugated, Clone: 3A7B2, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67088-1-IG-150UL
This item is not returnable.
View return policy
Description
The SDC2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human SDC2. This antibody recognizes human antigen. The SDC2 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
| SDC2 | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human |
| Western Blot, Immunohistochemistry, ELISA | |
| 3A7B2 | |
| Unconjugated | |
| 150 μL | |
| 6383 | |
| IgG2a |