Learn More
SCYL1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP2-94120-0.02ml
 View more versions of this product
Product Code. 18611972
  • 204.00€ / 0.02mL
Description
SCYL1 Polyclonal antibody specifically detects SCYL1 in Human, Mouse, Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SCYL1
Polyclonal
Western Blot 1:500 - 1:1000
Coated vesicle-associated kinase of 90 kDa, CVAK90, GKLPTelomerase transcriptional element-interacting factor, likely ortholog of mouse N-terminal kinase-like protein, MGC78454, NKTL, N-terminal kinase-like protein, NTKLN-terminal kinase-like, P105, SCY1-like 1 (S. cerevisiae), TAPKTeratoma-associated tyrosine kinase, TEIFSCY1-like protein 1, Telomerase regulation-associated protein, telomerase transcriptional elements-interacting factor, TRAPHT019
Recombinant fusion protein containing a sequence corresponding to amino acids 460-740 of human SCYL1 (NP_065731.3). SASTRHRVLTSAFSRATRDPFAPSRVAGVLGFAATHNLYSMNDCAQKILPVLCGLTVDPEKSVRDQAFKAIRSFLSKLESVSEDPTQLEEVEKDVHAASSPGMGGAAASWAGWAVTGVSSLTSKLIRSHPTTAPTETNIPQRPTPEGVPAPAPTPVPATPTTSGHWETQEEDKDTAEDSSTADRWDDEDWGSLEQEAESVLAQQDDWSTGGQVSRASQVSNSDHKSSKSPESDWSSWEAEGSWEQGWQEPSSQEPPPDGTRLASEYNWGGPESSDKGDPFA
0.02 mL
Protein Kinase
57410
Store at -20°C. Avoid freeze-thaw cycles.
IgG
Western Blot
Unconjugated
PBS (pH 7.3), 50% glycerol
Rabbit
Affinity purified
RUO
Primary
Human, Mouse, Rat