missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCUBE1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-77185-0.025mg
This item is not returnable.
View return policy
Description
SCUBE1 Polyclonal specifically detects SCUBE1 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| SCUBE1 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| signal peptide, CUB and EGF-like domain-containing protein 1, signal peptide, CUB domain, EGF-like 1, signal peptide-CUB domain-EGF-related 1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 80274 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| 1 mg/mL | |
| Western Blot 1 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 20 μg/mL | |
| NP_766638 | |
| SCUBE1 | |
| Antibody was raised against a 14 amino acid synthetic peptide near the center of human SCUBE1. The immunogen is located within amino acids 460 - 510 of SCUBE1. Amino Acid Squence: CSDAPTTPIKQKAR | |
| 0.025 mg | |
| Primary | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction