missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCP1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA131221
This item is not returnable.
View return policy
Description
PA1-31221 detects SCP1 from mouse, human samples. PA1-31221 has been successfully used in immunofluorescence applications. The PA1-31221 immunogen is: Synthetic peptide (Human) (C terminal).
The synaptonemal complex is a proteinaceous complex that apparently mediates synapses during the zygotene stage and then disintegrates. It is a complex structure that unites homologous chromosomes during the prophase stage of meiosis. It is the result of
Specifications
| SCP1 | |
| Polyclonal | |
| Unconjugated | |
| Q15431, Q62209 | |
| SYCP1 | |
| Synthetic peptide (Human) (C terminal). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| Immunocytochemistry, Immunofluorescence | |
| 1 mg/mL | |
| SYCP1 | |
| CT8, HOM-TES-14, MGC104417, SCP1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 20957, 6847 | |
| 4° C, do not freeze | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction