missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCP1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 45 publications
Brand: Novus Biologicals NB300-229
This item is not returnable.
View return policy
Description
SCP1 Polyclonal specifically detects SCP1 in Mouse, Rat, Chicken, Mammal, Parasite, Monkey, Human (Negative) samples. It is validated for Western Blot, Simple Western, Chromatin Immunoprecipitation, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
| SCP1 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| SYCP1 | |
| A synthetic peptide made to the C-terminus of the mouse SCP1 protein sequence. [UniProt# Q62209] | |
| 0.1 mL | |
| Cell Cycle and Replication, Chromatin Research, Growth and Development, Neuronal Cell Markers, Neuroscience, Neurotransmission | |
| 6847 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, ChIP Assay, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| 1.0 mg/mL | |
| Western Blot, Simple Western 1:100, Chromatin Immunoprecipitation 1:10 - 1:500. Use reported in scientific literature (PMID 27486799), Immunohistochemistry 1:750, Immunocytochemistry/Immunofluorescence 1:100 - 1:750, Immunoprecipitation, Immunohistochemistry-Paraffin 1:750, Immunohistochemistry-Frozen 1:750, Chromatin Immunoprecipitation (ChIP) 1:10-1:500 | |
| Cancer/testis antigen 8, CT8MGC104417, HOM-TES-14, SCP1SCP-1, synaptonemal complex protein 1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Mouse, Rat, Chicken, Mammalia, Parasite, Monkey, Human (Negative) | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction