missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCN5A Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 68273-1-IG-20UL
This item is not returnable.
View return policy
Description
The SCN5A antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human SCN5A. This antibody recognizes human, mouse, rat, rabbit antigen. The SCN5A antibody has been validated for the following applications: WB, IHC, FC (Intra), ELISA analysis.Specifications
| SCN5A | |
| Monoclonal | |
| 1000 μg/mL | |
| CDCD2; CMD1E; CMPD2; HB1; HB2; HBBD; HH1; ICCD; IVF; LQT3; Nav1.5; PFHB1; SSS1 | |
| Recombinant Protein | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat, Rabbit | |
| 227 kDa | |
| Liquid |
| Western Blot, Immunohistochemistry, Flow Cytometry (Intracellular Staining), ELISA | |
| 1C2B3 | |
| Unconjugated | |
| Mouse | |
| Protein G purification | |
| RUO | |
| 6331 | |
| Store at -20°C. Stable for one year after shipment. | |
| Monoclonal Antibody | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction