missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ SCL/Tal1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-29594
This item is not returnable.
View return policy
Description
SCL/Tal1 Polyclonal antibody specifically detects SCL/Tal1 in Human samples. It is validated for Immunoprecipitation,Western Blot
Specifications
| SCL/Tal1 | |
| Polyclonal | |
| Tris-citrate/phosphate pH 7 -8. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| BHLHA17, bHLHa17tal-1, Class A basic helix-loop-helix protein 17, SCLT-cell acute lymphocytic leukemia protein 1, Stem cell protein, TAL-1, T-cell acute lymphocytic leukemia 1, T-cell leukemia/lymphoma protein 5, TCL5tal-1 product | |
| Between 1 and 50 | |
| 100 μg | |
| Phospho Specific | |
| 6886 | |
| Store at 4°C. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction