missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCARF1 Rabbit Polyclonal Antibody, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 28174-1-AP-20UL
This item is not returnable.
View return policy
Description
The SCARF1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human SCARF1. This antibody recognizes human antigen. The SCARF1 antibody has been validated for the following applications: WB, ELISA analysis.Specifications
| SCARF1 | |
| Polyclonal | |
| Unconjugated | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 8578 | |
| Store at -20°C. Stable for one year after shipment. | |
| Polyclonal Antibody | |
| IgG |
| Western Blot, ELISA | |
| 500 μg/mL | |
| KIAA0149; MGC47738; SREC | |
| Recombinant Protein | |
| 20 μL | |
| Primary | |
| Human | |
| 90 kDa, 180 kDa | |
| Liquid |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?