missing translation for 'onlineSavingsMsg'
Learn More

SBSN Antibody (SBSN/7961), Alexa Fluor™ 488, Novus Biologicals™

Product Code. 18517489 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18517489 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18517489 Supplier Novus Biologicals Supplier No. NBP324300AF488

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

SBSN Monoclonal antibody specifically detects SBSN in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen SBSN
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone SBSN/7961
Conjugate Alexa Fluor 488
Formulation 50mM Sodium Borate
Gene Alias HLAR698, MGC75533, suprabasin, UNQ698
Host Species Mouse
Immunogen Recombinant fragment of human SBSN protein (exact sequence is proprietary)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 374897
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.