missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
SATB2 Monoclonal antibody specifically detects SATB2 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | SATB2 |
| Applications | Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | CL0276 |
| Conjugate | Janelia Fluor 525 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | DNA-binding protein SATB2, FLJ32076, KIAA1034FLJ21474, MGC119474, MGC119477, SATB family member 2, SATB homeobox 2, Special AT-rich sequence-binding protein 2 |
| Host Species | Mouse |
| Immunogen | This antibody was developed using a recombinant protein derived from Q9UPW6, with the exact immunogen sequence remaining proprietary. |
| Purification Method | Protein A purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?