missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SART1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
200.00€ - 450.00€
Specifications
| Antigen | SART1 |
|---|---|
| Concentration | 1.3 mg/mL |
| Dilution | Western Blot 1 ug/ml, Immunohistochemistry 1:200-1:500, Immunohistochemistry-Paraffin 1:200-1:500, ICC/IF (Negative) |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunocytochemistry |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18109051
|
Novus Biologicals
NBP2-14836SS |
0.025 mL |
200.00€
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18117581
|
Novus Biologicals
NBP2-14836 |
0.1 mL |
450.00€
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SART1 Polyclonal specifically detects SART1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, ICC/IF (Negative).Specifications
| SART1 | |
| Western Blot 1 ug/ml, Immunohistochemistry 1:200-1:500, Immunohistochemistry-Paraffin 1:200-1:500, ICC/IF (Negative) | |
| Polyclonal | |
| Rabbit | |
| Hypoxia | |
| PBS and 30% Glycerol with 0.05% Sodium Azide | |
| Allergen Hom s 1, Ara1, HAF, HOMS1, hSART-1, hSnu66, hypoxia-associated factor, IgE autoantigen, MGC2038, SART-1, SART1(259) protein, SART1(800) protein, SART1259, small nuclear ribonucleoprotein 110kDa (U4/U6.U5), SNRNP110, Snu66, SNU66 homolog, squamous cell carcinoma antigen recognised by T cells, squamous cell carcinoma antigen recognized by T cells, Squamous cell carcinoma antigen recognized by T cells 1, U4/U6.U5 tri-snRNP-associated 110 kDa protein, U4/U6.U5 tri-snRNP-associated protein 1 | |
| SART1 | |
| IgG | |
| Affinity Purified | |
| Human and mouse. Immunogen sequence has 94% homology to rat. |
| 1.3 mg/mL | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunocytochemistry | |
| Unconjugated | |
| RUO | |
| Human, Mouse | |
| O43290, Q9Z315 | |
| 9092 | |
| A synthetic peptide made to a internal portion of the human SART1 protein (between residues 100-200) [UniProt O43290] | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title