missing translation for 'onlineSavingsMsg'
Learn More

SARS-CoV-2 Spike RBD Antibody, R&D Systems™
GREENER_CHOICE

Mouse Monoclonal Antibody

214.00€ - 453.00€

Specifications

Antigen SARS-CoV-2 Spike RBD
Clone 1035753
Dilution ELISA, Blockade of Receptor-ligand Interaction 50 ug/mL
Applications ELISA, Blocking Assay
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18720817
GREENER_CHOICE
missing translation for 'viewDocuments'
R&D Systems
MAB105808-SP
25 μg
214.00€
25µg
View alternative products
missing translation for 'toSaveUpto' 
»
Please to purchase this item. Need a web account? Register with us today!
18744197
GREENER_CHOICE
missing translation for 'viewDocuments'
R&D Systems
MAB105808-100
100 μg
453.00€
100µg
View alternative products
missing translation for 'toSaveUpto' 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Spike RBD Monoclonal specifically detects Spike RBD in SARS-CoV-2 samples. It is validated for ELISA, Blockade of Receptor-ligand Interaction.
Specifications

Specifications

SARS-CoV-2 Spike RBD
ELISA, Blockade of Receptor-ligand Interaction 50 ug/mL
Monoclonal
Purified
RUO
Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS.
2019-nCoV Peplomer protein, 2019-nCoV S Protein, 2019-nCoV Spike, COVID-19 Spike, E2, Human coronavirus spike glycoprotein, Peplomer protein, S glycoprotein, S protein, SARS-CoV-2, SARS-COV-2 E2, SARS-COV-2 Peplomer protein, SARS-COV-2 S protein, SARS-COV-2 Spike glycoprotein, SARSCOV2 Spike protein, Severe Acute Respiratory Syndrome Coronavirus 2 Spike Protein, Spike, Spike glycoprotein, Spike S1, Spike S1 RBD, surface glycoprotein
Spodoptera frugiperda, Sf 21 (baculovirus)-derived SARS-CoV-2 Spike S1 Subunit, Val16-Pro681, Accession # YP_009724390.1
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
1035753
ELISA, Blocking Assay
Unconjugated
Mouse
SARS-CoV-2
YP_009724390.1
43740568
IgG1
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Videos
SDS
Documents

Documents

Certificates
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.