Learn More
Invitrogen™ SARS-CoV-2 Spike Protein RBD Omicron (XBB.1) Chimeric Recombinant Human Monoclonal Antibody (D008)
Human Recombinant Monoclonal Antibody
279.00€ - 569.00€
Specifications
| Antigen | SARS-CoV-2 Spike Protein RBD Omicron (XBB.1) Chimeric |
|---|---|
| Clone | D008 |
| Concentration | 1 mg/mL |
| Content And Storage | -20°C, Avoid Freeze/Thaw Cycles |
| Applications | ELISA |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30535638
|
Invitrogen™
MA560610 |
100 μg |
569.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30535322
|
Invitrogen™
MA560611 |
20 μg |
279.00€
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Specifically recognize the SARS-CoV-2 Omicron XBB.1 and XBB Spike RBD and ECD. No cross-reactivity with SARS-CoV-2 BF.7, BA.4/BA.5/BA.5.2, BA.2.75, BQ.1.1, wild type, Alpha, Beta, Delta. Minimal cross-reactivity with SARS-CoV-2 XBB.1.5.. Has cross-reactivity in ELISA with: SARS-CoV-2 Omicron (XBB.1) Spike S1+S2 trimer Protein; SARS-CoV-2 Omicron (XBB) Spike S1+S2 trimer Protein; SARS-CoV-2 Omicron (XBB) Spike RBD Protein; Minimal cross-reactivity in ELISA with: SARS-CoV-2 Omicron (XBB.1.5) Spike S1+S2 trimer Protein; SARS-CoV-2 Omicron (XBB.1.5) Spike RBD Protein; No cross-reactivity in ELISA with: SARS-CoV-2 Omicron (BF.7) Spike S1+S2 trimer Protein; SARS-CoV-2 Omicron (BA.4.6/BF.7) Spike RBD Protein; SARS-CoV-2 Omicron (BA.4/BA.5/BA.5.2) Spike S1+S2 trimer Protein; SARS-CoV-2 Omicron (BA.4/BA.5/BA.5.2) Spike RBD Protein; SARS-CoV-2 Omicron (B.1.1.529) S1+S2 trimer Protein; SARS-CoV-2 Omicron (B.1.1.529) Spike RBD Protein; SARS-CoV-2 Omicron (BQ.1.1) Spike S1+S2 trimer Protein; SARS-CoV-2 Omicron (BQ.1.1) Spike RBD Protein; SARS-CoV-2 Omicron (BA.2.75.2) Spike S1+S2 trimer Protein; SARS-CoV-2 Omicron (BA.2.75) Spike RBD Protein; SARS-CoV-2 Delta (B.1.617.2) Spike S1+S2 trimer Protein; SARS-CoV-2 Delta (B.1.617.2) Spike RBD Protein; SARS-CoV-2 Beta (B.1.351) Spike S1+S2 trimer Protein; SARS-CoV-2 Beta (B.1.351, B.1.351.2, B.1.351.3) Spike RBD Protein; SARS-CoV-2 Alpha (B.1.1.7) Spike S1+S2 trimer Protein; SARS-CoV-2 Alpha (B.1.1.7) Spike RBD Protein; SARS-CoV-2 Spike S1+S2...
The severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2) variant Omicron (B 1 1 529) has attracted great concerns since its identification in South Africa Omicron is the fifth variant of concern (VOC) after Alpha (B 1 1 7), Beta (B 1 351), Gamma (P 1) and Delta (B 1 617 2) Omicron variant harbors >30 substitutions/deletions/insertions in the spike, including 15 substitutions in receptor-binding domain (RBD) [PMID: 35397623].Specifications
| SARS-CoV-2 Spike Protein RBD Omicron (XBB.1) Chimeric | |
| 1 mg/mL | |
| ELISA | |
| Unconjugated | |
| Human | |
| RUO | |
| PBS with no preservative | |
| Recombinant SARS-CoV-2 Omicron XBB.1 Spike ECD Protein. | |
| Antibody |
| D008 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG1 κ | |
| Virus | |
| 2019-nCoV Spike; COVID-19 Spike; SARS-CoV-2 Spike | |
| Primary | |
| Protein A |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.