missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CES2 Polyclonal Antibody
GREENER_CHOICE

Goat Polyclonal Antibody

Brand:  Invitrogen™ PA5149043

Product Code. 30535323

  • 469.00€ / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Reconstitute at 0.2 mg/mL in sterile PBS.

CES2 is a member of the carboxylesterase large family. The family members are responsible for the hydrolysis or transesterification of various xenobiotics, such as cocaine and heroin, and endogenous substrates with ester, thioester, or amide bonds. They may participate in fatty acyl and cholesterol ester metabolism, and may play a role in the blood-brain barrier system. The protein encoded by this gene is the major intestinal enzyme and functions in intestine drug clearance.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CES2
Polyclonal
Unconjugated
CES2
9030624L02Rik; 9130231C15Rik; ACH M1; Acylcarnitine hydrolase; acylcarnitine hydrolase M1; AI266984; Carboxylesterase 2; carboxylesterase 2 (intestine, liver); carboxylesterase 2A; carboxylesterase 2C; carboxylesterase 2E; carboxylesterase 5; carboxylesterase 6; carboxylesterase isoenzyme; carboxylesterase ML3; CE-2; CES 2; Ces2; CES-2; Ces2a; CES2A1; ces2A3; Ces2c; Ces2e; Ces2l; Ces5; Ces6; CESRL4; Cocaine esterase; hCE-2; ICE; intestinal carboxylesterase; liver carboxylesterase-2; methylumbelliferyl-acetate deacetylase 2; P iCE; PCE-2; Peroxisome proliferator-inducible acylcarnitine hydrolase; phenobarbital-inducible carboxylesterase (liver); pyrethroid hydrolase Ces2a; Pyrethroid hydrolase Ces2e
Goat
Antigen affinity chromatography
RUO
8824
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Immunohistochemistry (Paraffin), Western Blot
0.2 mg/mL
PBS with trehalose and no preservative
O00748
CES2
Mouse myeloma cell line NS0-derived recombinant human Carboxylesterase 2/CES2, Gln27-Leu559.
100 μg
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.