Learn More
ACROBiosystems SARS-CoV-2 S protein RBD (N501Y), Fc Tag
Brand:  ACROBiosystems SPD-C5253-100ug
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 17206510
  • 522.57€ / 100µg
Estimated Shipment: 07-08-2024
to see stock.
Add to basket
Description
Description
It's been reported that Coronavirus can infect the human respiratory epithelial cells through interaction with the human ACE2 receptor. The spike protein is a large type I transmembrane protein containing two subunits, S1 and S2. S1 mainly contains a receptor binding domain (RBD), which is responsible for recognizing the cell surface receptor. S2 contains basic elements needed for the membrane fusion.The S protein plays key parts in the induction of neutralizing-antibody and T-cell responses, as well as protective immunity.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 51.1 kDa.
Specifications
Specifications
51.1 kDa
SARS-CoV-2 S protein RBD (N501Y), Fc Tag (SPD-C5253) is expressed from human 293 cells (HEK293). It contains AA Arg 319 - Lys 537 (Accession # QHD43416.1 (N501Y)).
-20°C
S protein RBD
SARS-CoV-2
HEK293
Viral Proteins
100 μg
Arg 319 - Lys 537
Spike,S protein RBD,Spike glycoprotein Receptor-binding domain,S glycoprotein RBD,Spike protein RBD,COVID-19
Unconjugated
Fc Tag
Powder