missing translation for 'onlineSavingsMsg'
Learn More

SARS-CoV-2 S protein (RBD, 319-541 aa) Mouse Monoclonal Antibody, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 67758-1-IG-150UL

 View more versions of this product

Product Code. 30235957

  • ONLINE EXCLUSIVE PRICE
    410.00€
    349.00€ / 150µL
    Save 61.00€
    14% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The SARS-CoV-2 S protein (RBD, 319-541 aa) antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of virus SARS-CoV-2 S protein (RBD, 319-541 aa). This antibody recognizes virus antigen. The SARS-CoV-2 S protein (RBD, 319-541 aa) antibody has been validated for the following applications: WB, ELISA, Neutralization analysis.
Specifications

Specifications

SARS-CoV-2 S protein (RBD, 319-541 aa)
Monoclonal
1000 μg/mL
Mouse
Protein A purification
RUO
43740568
Store at -20°C. Stable for one year after shipment.
Liquid
Western Blot, ELISA, Neutralization
1H3E9
Unconjugated
Recombinant Protein
150 μL
Primary
Virus
Monoclonal Antibody
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.