Learn More
IBA Lifesciences SARS-CoV-2 Peptide LSPRWYFYY (HLA-A*2402)
GREENER_CHOICE
Product Code. 16644754 Shop All IBA Lifesciences Products
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Product Code. 16644754
Brand: IBA Lifesciences 67123901
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Single peptide synthesized as presented by the MHC class I allele
  • SARS-CoV-2 peptide LSPRWYFYY (HLA-A*2402) is a single peptide for stimulation of T cells.
  • The peptide from SARS-CoV-2 is synthesized as it is presented by the MHC class I HLA-A*2402 allele and can be used for targeted in vitro generation and expansion of antigen-specific CD8+ T cells.
  • The peptides were purified by HPLC using a water and acetonitrile gradient and are endotoxin-free.
Specifications
Purity >85%
Endotoxin Level Free
Format Lyophilized Powder
Quantity 1 mg
Research Category T cell stimulation
For Research Use Only.