Learn More
Invitrogen™ SARS-CoV-2 Omicron Spike Protein S1/S2 (XBB.1) Chimeric Recombinant Human Monoclonal Antibody (D007)
GREENER_CHOICE
Product Code. 30534339
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µg
20µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30534339 100 μg 100µg
30535325 20 μg 20µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30534339 Supplier Invitrogen™ Supplier No. MA560608
Human Recombinant Monoclonal Antibody
Specifically recognize the SARS-CoV-2 Omicron XBB.1 Spike. No cross-reactivity with SARS-CoV-2 XBB.1.5, XBB, BF.7, BA.4/BA.5/BA.5.2, BA.2.75, BQ.1.1, wild type, Alpha, Beta, Delta.. Has cross-reactivity in ELISA with: SARS-CoV-2 Omicron (XBB.1) Spike S1+S2 trimer Protein; No cross-reactivity in ELISA with: SARS-CoV-2 Omicron (XBB.1.5) Spike S1+S2 trimer Protein; SARS-CoV-2 Omicron (XBB.1.5) Spike RBD Protein; SARS-CoV-2 Omicron (XBB) Spike S1+S2 trimer Protein; SARS-CoV-2 Omicron (XBB) Spike RBD Protein; SARS-CoV-2 Omicron (BF.7) Spike S1+S2 trimer Protein; SARS-CoV-2 Omicron (BA.4.6/BF.7) Spike RBD Protein; SARS-CoV-2 Omicron (BA.4/BA.5/BA.5.2) Spike S1+S2 trimer Protein; SARS-CoV-2 Omicron (BA.4/BA.5/BA.5.2) Spike RBD Protein; SARS-CoV-2 Omicron (B.1.1.529) S1+S2 trimer Protein; SARS-CoV-2 Omicron (B.1.1.529) Spike RBD Protein; SARS-CoV-2 Omicron (BQ.1.1) Spike S1+S2 trimer Protein; SARS-CoV-2 Omicron (BQ.1.1) Spike RBD Protein; SARS-CoV-2 Omicron (BA.2.75.2) Spike S1+S2 trimer Protein; SARS-CoV-2 Omicron (BA.2.75) Spike RBD Protein; SARS-CoV-2 Delta (B.1.617.2) Spike S1+S2 trimer Protein; SARS-CoV-2 Delta (B.1.617.2) Spike RBD Protein; SARS-CoV-2 Beta (B.1.351) Spike S1+S2 trimer Protein; SARS-CoV-2 Beta (B.1.351, B.1.351.2, B.1.351.3) Spike RBD Protein; SARS-CoV-2 Alpha (B.1.1.7) Spike S1+S2 trimer Protein; SARS-CoV-2 Alpha (B.1.1.7) Spike RBD Protein; SARS-CoV-2 Spike S1+S2 (D614G) trimer Protein; SARS-CoV-2 Spike RBD Protein.
The SARS-CoV-2 Omicron Spike Protein S1/S2 refers to specific regions of the spike (S) protein of the Omicron variant of the SARS-CoV-2 virus, which is responsible for COVID-19. The spike protein is a key component of the virus's structure and plays a critical role in its ability to infect host cells. It is composed of two main subunits: S1 and S2. The S1/S2 designation refers to the cleavage site between these two subunits. This cleavage is a crucial step for the activation of the spike protein, which is necessary for the virus to enter host cells. The cleavage is typically performed by host cell proteases, such as furin. The Omicron variant of SARS-CoV-2 has numerous mutations in the spike protein, including in the S1 and S2 subunits. These mutations can affect the virus's transmissibility, ability to evade the immune response, and the effectiveness of vaccines and treatments.
TRUSTED_SUSTAINABILITY
Specifications
Antigen SARS-CoV-2 Omicron Spike Protein S1/S2 (XBB.1) Chimeric
Applications ELISA
Classification Recombinant Monoclonal
Clone D007
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Host Species Human
Immunogen Recombinant SARS-CoV-2 Omicron XBB.1 Spike ECD Protein.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1 κ