missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SARS-CoV-2 Nucleocapsid Antibody (004), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-12734-100ul
This item is not returnable.
View return policy
Description
SARS-CoV-2 Nucleocapsid Monoclonal antibody specifically detects SARS-CoV-2 Nucleocapsid in SARS-CoV samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| SARS-CoV-2 Nucleocapsid | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| SARS-CoV | |
| Purified |
| Western Blot, Flow Cytometry, ELISA, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 004 | |
| Western Blot 1:1000-1:10000, Flow Cytometry 1:25-1:100, ELISA 1:5000-1:10000, Immunocytochemistry/ Immunofluorescence 1:20-1:100, Immunohistochemistry-Paraffin 1:100-1:500 | |
| 2019-nCoV N Protein, 2019-nCoV Nucleocapsid Protein, COVID-19 N protein, COVID-19 nucleocapsid protein, Human coronavirus Nucleoprotein, nucleocapsid phosphoprotein, Nucleocapsid protein, Nucleoprotein, ORF9; structural protein, SARS-CoV-2, SARS-CoV-2 Nucleocapsid protein, SARS-CoV-2 N protein, SARSCoV2 Nucleoprotein, Severe Acute Respiratory Syndrome Coronavirus 2 Nucleocapsid Protein | |
| Recombinant SARS-CoV-2 Nucleocapsid Protein | |
| 100 μg | |
| Virology Bacteria and Parasites | |
| 43740575 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction