missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SARS-CoV-2 Nucleocapsid (aa1-419), His Tag Recombinant Protein
HEK293 cells Recombinant Protein
564.00€ - 3605.00€
Specifications
For Use With (Application) | Control, ELISA, Western Blot |
---|---|
Formulation | PBS with no preservative |
Name | SARS-CoV-2 Nucleocapsid (aa1-419), His Tag |
Source | HEK293 cells |
Storage Requirements | -20°Cor to +80°C if preferred |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17194599
|
Invitrogen™
RP87690 |
1 mg |
3605.00€
1mg |
Estimated Shipment: 05-06-2024
Log in to see stock. |
|||||
17104599
|
Invitrogen™
RP87676 |
100 μg |
780.00€
100µg |
Estimated Shipment: 05-06-2024
Log in to see stock. |
|||||
17174599
|
Invitrogen™
RP87688 |
50 μg |
564.00€
50µg |
Estimated Shipment: 05-06-2024
Log in to see stock. |
|||||
17184599
|
Invitrogen™
RP87689 |
200 μg |
1260.00€
200µg |
Estimated Shipment: 05-06-2024
Log in to see stock. |
|||||
Description
Severe Acute Respiratory Syndrome (SARS), an emerging disease characterized by atypical pneumonia, has recently been attributed to a novel coronavirus (SARS-CoV). The nucleocapsid protein (N) was one of the structural proteins of SARS-CoV (N, S, sE, M). The severe acute respiratory syndrome coronavirus (SARS-CoV) nucleocapsid (N) protein is the most abundant protein in the virus-infected cells. The SARS-CoV-2 S gene has 76% amino acid similarity to the SARS-CoV S gene, and nonsynonymous mutations developed in the S protein as the SARS-CoV epidemic developed, reports have shown that the crystal structure of the SARS-CoV-2 nucleocapsid protein is similar to those of previously described coronavirus N proteins, but their surface electrostatic potential characteristics are distinct.Specifications
Control, ELISA, Western Blot | |
SARS-CoV-2 Nucleocapsid (aa1-419), His Tag | |
-20°Cor to +80°C if preferred | |
Coronavirus; COVID; COVID 19; COVID19; COVID-19; SARS; SARS Coronavirus; SARS CoV; SARS CoV2; SARS CoV-2; SARS Nucleocapsid; SARS-CoV2; SARS-CoV-2 | |
Unconjugated | |
Lyophilized |
PBS with no preservative | |
HEK293 cells | |
RUO | |
Virus | |
Recombinant | |
>95% |