missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SARS-CoV-2 Nucleocapsid (aa1-419), His Tag Recombinant Protein

HEK293 cells Recombinant Protein

Brand:  Invitrogen™ RP87690

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 17194599

  • 3605.00€ / 1mg
Estimated Shipment: 20-06-2024
to see stock.
Add to basket

Description

Description

Severe Acute Respiratory Syndrome (SARS), an emerging disease characterized by atypical pneumonia, has recently been attributed to a novel coronavirus (SARS-CoV). The nucleocapsid protein (N) was one of the structural proteins of SARS-CoV (N, S, sE, M). The severe acute respiratory syndrome coronavirus (SARS-CoV) nucleocapsid (N) protein is the most abundant protein in the virus-infected cells. The SARS-CoV-2 S gene has 76% amino acid similarity to the SARS-CoV S gene, and nonsynonymous mutations developed in the S protein as the SARS-CoV epidemic developed, reports have shown that the crystal structure of the SARS-CoV-2 nucleocapsid protein is similar to those of previously described coronavirus N proteins, but their surface electrostatic potential characteristics are distinct.
Specifications

Specifications

Control, ELISA, Western Blot
SARS-CoV-2 Nucleocapsid (aa1-419), His Tag
HEK293 cells
RUO
Virus
Recombinant
>95%
PBS with no preservative
1 mg
-20°Cor to +80°C if preferred
Coronavirus; COVID; COVID 19; COVID19; COVID-19; SARS; SARS Coronavirus; SARS CoV; SARS CoV2; SARS CoV-2; SARS Nucleocapsid; SARS-CoV2; SARS-CoV-2
Unconjugated
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers