missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SARS-CoV-2 nsp13 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-07056CL1
This item is not returnable.
View return policy
Description
SARS-CoV-2 nsp13 Polyclonal antibody specifically detects SARS-CoV-2 nsp13 in SARS-CoV-2 samples. It is validated for ELISA,Immunohistochemistry
Specifications
| SARS-CoV-2 nsp13 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| SARS-CoV-2 | |
| Purified |
| ELISA, Immunohistochemistry | |
| CoraFluor 1 | |
| helicase, Non-structural protein 13 | |
| This antibody was raised against a peptide corresponding to 14 amino acids near the center of the SARS-CoV-2 (COVID-19) NSP13 (Helicase) protein. The immunogen is located between 440-490 amino acids of SARS-CoV-2 (COVID-19) NSP13 (Helicase). | |
| 0.1 mL | |
| Infections (Virus Bacteria and Parasites) | |
| 43740578 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction