missing translation for 'onlineSavingsMsg'
Learn More

SARS-COV-2 NSP1 Rabbit Polyclonal Antibody, Proteintech

Rabbit Polyclonal Antibody

145.00€ - 410.00€

Specifications

Antigen SARS-COV-2 NSP1
Concentration 1000 μg/mL
Applications ELISA
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30236011
View Documents
Proteintech
29287-1-AP-20UL
20 μL
145.00€
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30237877
View Documents
Proteintech
29287-1-AP-150UL
150 μL
410.00€
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The SARS-COV-2 NSP1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of sars-cov-2 SARS-COV-2 NSP1. This antibody recognizes virus antigen. The SARS-COV-2 NSP1 antibody has been validated for the following applications: ELISA analysis.
Specifications

Specifications

SARS-COV-2 NSP1
ELISA
Unconjugated
Rabbit
Virus
Recombinant Protein
Primary
Store at -20°C. Stable for one year after shipment.
1000 μg/mL
Polyclonal
Liquid
RUO
43740578
IgG
Antigen affinity purification
Polyclonal Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.