Learn More
Invitrogen™ SARS-CoV-2 Delta Spike Protein RBD Chimeric Recombinant Human Monoclonal Antibody (D006)
GREENER_CHOICE
Product Code. 30534332
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µg
20µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30534332 100 μg 100µg
30534319 20 μg 20µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30534332 Supplier Invitrogen™ Supplier No. MA560606
Human Recombinant Monoclonal Antibody
Specifically recognize the SARS-CoV-2 Delta Spike RBD and ECD. No cross-reactivity with SARS-CoV-2 wild type, Alpha, Beta, Omicron BA.2.75 variant.. Has cross-reactivity in ELISA with: SARS-CoV-2 Delta (B.1.617.2) Spike S1+S2 trimer Protein; SARS-CoV-2 Delta (B.1.617.2) Spike RBD Protein; No cross-reactivity in ELISA with: SARS-CoV-2 Omicron (XBB.1.16) Spike S1+S2 trimer Protein; SARS-CoV-2 Omicron (XBB.1.5) Spike RBD Protein; SARS-CoV-2 Omicron (XBB.1.5) Spike S1+S2 trimer Protein; SARS-CoV-2 Omicron (XBB.1) Spike S1+S2 trimer Protein; SARS-CoV-2 Omicron (XBB) Spike RBD Protein; SARS-CoV-2 Omicron (BQ.1.1) Spike S1+S2 trimer Protein; SARS-CoV-2 Omicron (BQ.1.1)Spike RBD Protein; SARS-CoV-2 Omicron (BF.7) Spike S1+S2 trimer Protein; SARS-CoV-2 Omicron (BA.4.6/BF.7) Spike RBD Protein; SARS-CoV-2 Omicron (BA.4.6) Spike S1+S2 trimer Protein; SARS-CoV-2 Omicron (BA.2.75) Spike S1+S2 trimer Protein; SARS-CoV-2 Omicron (BA.2.75.2) Spike RBD Protein; SARS-CoV-2 Omicron (BA.2.75.2) Spike S1+S2 trimer Protein; SARS-CoV-2 Omicron (BA.4/BA.5/BA.5.2) Spike S1+S2 trimer Protein; SARS-CoV-2 Omicron (BA.4/BA.5/BA.5.2) Spike RBD Protein; SARS-CoV-2 Omicron (BA.2.75) Spike RBD Protein; SARS-CoV-2 Omicron (BA.2.3.20) Spike RBD Protein; SARS-CoV-2 Omicron (B.1.1.529) S1+S2 trimer Protein; SARS-CoV-2 Omicron (B.1.1.529) Spike RBD Protein; SARS-CoV-2 Alpha (B.1.1.7) Spike S1+S2 trimer Protein; SARS-CoV-2 Alpha (B.1.1.7) Spike RBD Protein; SARS-CoV-2 Beta (B.1.351) Spike S1+S2 trimer Protein; S...
The SARS-CoV-2 Delta Spike Protein RBD refers to the receptor-binding domain (RBD) of the spike (S) protein in the Delta variant of the SARS-CoV-2 virus, which is responsible for COVID-19. The RBD is a critical region within the S1 subunit of the spike protein that binds to the angiotensin-converting enzyme 2 (ACE2) receptor on human cells, facilitating viral entry. The Delta variant, first identified in India, possesses several mutations in the RBD that can enhance its binding affinity to the ACE2 receptor, potentially increasing the virus's transmissibility and partially evading immune responses. These mutations have significant implications for the effectiveness of vaccines and therapeutic antibodies, making it essential to monitor and adapt public health strategies accordingly.
TRUSTED_SUSTAINABILITY
Specifications
Antigen SARS-CoV-2 Delta Spike Protein RBD Chimeric
Applications ELISA, Neutralization
Classification Recombinant Monoclonal
Clone D006
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Host Species Human
Immunogen Recombinant SARS-CoV-2 Delta Spike ECD Protein.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1 κ