Learn More
Invitrogen™ SARS-CoV-2 Alpha Spike Protein RBD Chimeric Recombinant Human Monoclonal Antibody (D004)
GREENER_CHOICE
Product Code. 30535066
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µg
20µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30535066 100 μg 100µg
30534989 20 μg 20µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30535066 Supplier Invitrogen™ Supplier No. MA560602
Human Recombinant Monoclonal Antibody
Specifically recognize the SARS-CoV-2 Alpha Spike RBD and ECD. No cross-reactivity with SARS-CoV-2, Beta, Delta, Omicron BA.4/BA.5/BA.5.2, Omicron BA.2.75 variant. Has Medium cross-reactivity in ELISA with SARS-CoV-2 wild type Spike RBD.. Has cross-reactivity in ELISA with: SARS-CoV-2 Alpha (B.1.1.7) Spike S1+S2 trimer Protein; SARS-CoV-2 Alpha (B.1.1.7) Spike RBD Protein; Has Medium cross-reactivity in ELISA with: SARS-CoV-2 Spike RBD Protein; No cross-reactivity in ELISA with: SARS-CoV-2 Omicron (XBB.1.16) Spike S1+S2 trimer Protein; SARS-CoV-2 Omicron (XBB.1.5) Spike RBD Protein; SARS-CoV-2 Omicron (XBB.1.5) Spike S1+S2 trimer Protein; SARS-CoV-2 Omicron (XBB.1) Spike S1+S2 trimer Protein; SARS-CoV-2 Omicron (XBB) Spike RBD Protein; SARS-CoV-2 Omicron (BQ.1.1) Spike S1+S2 trimer Protein; SARS-CoV-2 Omicron (BQ.1.1) Spike RBD Protein; SARS-CoV-2 Omicron (BF.7) Spike S1+S2 trimer Protein; SARS-CoV-2 Omicron (BA.4.6/BF.7) Spike RBD Protein; SARS-CoV-2 Omicron (BA.4.6) Spike S1+S2 trimer Protein; SARS-CoV-2 Omicron (BA.4/BA.5/BA.5.2) Spike S1+S2 trimer Protein; SARS-CoV-2 Omicron (BA.4/BA.5/BA.5.2) Spike RBD Protein; SARS-CoV-2 Omicron (BA.2.75) Spike RBD Protein; SARS-CoV-2 Omicron (BA.2.75) Spike S1+S2 trimer Protein; SARS-CoV-2 Omicron (BA.2.75.2) Spike RBD Protein; SARS-CoV-2 Omicron (BA.2.75.2) Spike S1+S2 trimer Protein; SARS-CoV-2 Omicron (BA.2.75) Spike RBD Protein; SARS-CoV-2 Omicron (BA.2.3.20) Spike RBD Protein; SARS-CoV-2 Omicron (B.1.1.529) S1+S2 trimer Protein...
The SARS-CoV-2 Alpha Spike Protein RBD refers to the receptor-binding domain (RBD) of the spike (S) protein found in the Alpha variant of the SARS-CoV-2 virus, which is responsible for COVID-19. The RBD is a crucial region within the S1 subunit of the spike protein that specifically binds to the angiotensin-converting enzyme 2 (ACE2) receptor on the surface of human cells, facilitating viral entry. The Alpha variant, first identified in the UK, has several mutations in the spike protein, including in the RBD, which can enhance the virus's ability to bind to ACE2, potentially increasing its transmissibility and impacting the effectiveness of immune responses. Understanding these mutations is vital for developing and adjusting vaccines and therapeutic strategies.
TRUSTED_SUSTAINABILITY
Specifications
Antigen SARS-CoV-2 Alpha Spike Protein RBD Chimeric
Applications ELISA, Neutralization
Classification Recombinant Monoclonal
Clone D004
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Host Species Human
Immunogen Recombinant SARS-CoV-2 Alpha Spike ECD Protein.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1 κ