missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sarcalumenin Antibody (XIIC4), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
555.00€
Specifications
| Antigen | Sarcalumenin |
|---|---|
| Clone | XIIC4 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Conjugate | FITC |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18557251
|
Novus Biologicals
NB120-2874F |
0.1 mL | |||||||
Description
Sarcalumenin Monoclonal antibody specifically detects Sarcalumenin in Mouse, Canine, Guinea Pig, Rabbit samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
| Sarcalumenin | |
| Western Blot | |
| FITC | |
| Mouse | |
| Apoptosis, Cancer, Tumor Suppressors | |
| PBS | |
| 6345 | |
| IgG1 | |
| Protein A purified |
| XIIC4 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Mouse | |
| sarcalumenin | |
| Purified rabbit sarcoplasmic reticulum membrane vesicles. | |
| Primary | |
| Store at 4°C in the dark. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title