missing translation for 'onlineSavingsMsg'
Learn More

SAP30L Antibody, mFluor Violet 500 SE, Novus Biologicals™

Product Code. 18777731 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18777731 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18777731 Supplier Novus Biologicals Supplier No. NB100491MFV500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SAP30L Polyclonal antibody specifically detects SAP30L in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen SAP30L
Applications Western Blot
Classification Polyclonal
Conjugate mFluor Violet 500 SE
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias DKFZp667L2214, FLJ11526, FLJ36497, HCV non-structural protein 4A-transactivated protein 2, histone deacetylase complex subunit SAP30L, NS4ATP2FLJ23595, SAP30-like, Sin3 corepressor complex subunit SAP30L, Sin3A associated protein p30-like, Sin3-associated protein p30-like
Host Species Rabbit
Immunogen A synthetic peptide made to residues 165-183 (VKSNKSRLDQKSEGGKQLE) of human SAP30L.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79685
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.