missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SAP102 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
200.00€
Specifications
| Antigen | SAP102 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 3 μg/mL, Immunohistochemistry 1:10 - 1:500, Immunohistochemistry-Frozen 10 μg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
Description
SAP102 Polyclonal specifically detects SAP102 in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Frozen.Specifications
| SAP102 | |
| Western Blot 3 μg/mL, Immunohistochemistry 1:10 - 1:500, Immunohistochemistry-Frozen 10 μg/mL | |
| Polyclonal | |
| Rabbit | |
| Cell Cycle and Replication, Neuroscience | |
| PBS with 1 mg/ml BSA with 0.05% Sodium Azide | |
| 1741 | |
| Synthetic peptide corresponding to residues G(83) P V P P K P V P G K N T P K L N G(100) of rat SAP102. | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Mouse | |
| discs, large homolog 3 (Drosophila), KIAA1232discs, large homolog 3 (neuroendocrine-dlg, Drosophila), MRX90MRX, NEDLGdisks large homolog 3, neuroendocrine-DLG, SAP-102, SAP102NE-Dlg, Synapse-associated protein 102, XLMR | |
| DLG3 | |
| IgG | |
| Affinity Purified | |
| SAP102 |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title