Learn More
SAMHD1 Antibody (OTI3F5), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 3 publications
Brand: Novus Biologicals NBP2-03285
This item is not returnable.
View return policy
Description
SAMHD1 Monoclonal specifically detects SAMHD1 in Human, Mouse, Rat, Canine, Primate, Monkey samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| SAMHD1 | |
| Human, Mouse | |
| IgG2b |
| Affinity Purified | |
| Store at -20°C. Avoid freeze-thaw cycles. |