missing translation for 'onlineSavingsMsg'
Learn More

Abcam Salmeterol Xinafoate, Beta2 Adrenoceptor Agonist, Solid, 50 mg

Product Code. 32110590 Shop All Abcam Products
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32110590 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32110590 Supplier Abcam Supplier No. ab12077150mg
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Selective, long acting β2 adrenoceptor agonist that inhibits the release of histamine and leukotrienes.

  • Molecular weight: 603.7 Da
  • Purity: >96%
  • Selective, long acting β2 adrenoceptor agonist
  • Inhibits release of histamine, leukotrienes, prostaglandin D2 and GM-CSF in a concentration-dependent manner
  • Active in vitro and in vivo

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Content And Storage Store at ambient temperature.
Form Solid
Shipping Condition Ambient
Target Molecule Beta2 Adrenoceptor
Quantity 50 mg
For Use With (Application) Sample Prep and Detection
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.