missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Salivary Amylase Alpha Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35327-100ul
This item is not returnable.
View return policy
Description
Salivary Amylase Alpha Polyclonal antibody specifically detects Salivary Amylase Alpha in Mouse samples. It is validated for ELISA,Western Blot
Specifications
| Salivary Amylase Alpha | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| 1,4-alpha-D-glucan glucanohydrolase 1, alpha-amylase 1, AMY1, AMY1B, AMY1C, amylase, alpha 1A (salivary), amylase, alpha 1A; salivary, amylase, salivary, alpha-1A, EC 3.2.1.1, glycogenase, Salivary alpha-amylase, salivary amylase alpha 1A | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 16-200 of human Salivary Amylase Alpha (NP_004029.2).,, Sequence:, QYSSNTQQGRTSIVHLFEWRWVDIALECERYLAPKGFGGVQVSPPNENVAIHNPFRPWWERYQPVSYKLCTRSGNEDEFRNMVTRCNNVGVRIYVDAVINHMCGNAVSAGTSSTCGSYFNPGSRDFPAVPYSGWDFNDGKCKTGSGDIENYNDATQVRDCRLSGLLDLALGKDYVRSKIAEYMNH | |
| 100 μL | |
| Primary | |
| Mouse | |
| Purified |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 276 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction