Learn More
Saline-Sodium Citrate (SSC), 20X Solution (Molecular Biology), Fisher BioReagents
Product Code. 10418353
Change view
Click to view available options
Quantity:
1L
4L
20L
Unit Size:
1 L
20 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10418353 1L 1 L
10785104 20L 20 L
10183433 4L 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10418353 Supplier Fisher BioReagents Supplier No. BP13251
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Saline–Sodium Citrate (SSC) is used as a buffer in Southern transfer protocols.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Saline-Sodium Citrate
Concentration 2.9 to 3.1 M
pH 7
Physical Form Liquid
Quantity 1L
DNase DNase free
Grade Molecular Biology