Learn More
Pro-Lab Diagnostics™ Safranin Stain
Product Code. 12871813
Click to view available options
Quantity:
500mL
1 L
Unit Size:
1 L
500mL
Product Code. 12871813
Brand: Pro-Lab Diagnostics™ PL.7012
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Use for Gram staining of bacteria. Pro-Lab Diagnostics™ Safranin Stain is designed to be ready to use.
  • Pro-Lab Diagnostics offers an extensive range of ready-to-use and concentrated microbiological staining solutions in a selection of convenient and economical sizes.
  • Manufactured in accordance with all current regulatory requirements
  • CE labelled
  • Strict quality control and filtering techniques prior to release
  • Also available are a range of convenient staining kits for Gram, TB and Cryptosporidium staining
Specifications
Solution Type Reagent
Type Safranin
Quantity 500mL
Certifications/Compliance (EC) No 1907/2006, Annex II, (EU) No 453/2010, CE-mark
Format Liquid
For Use With (Application) For Gram staining
Color Dark Red
Boiling Point 78 to 100°C
Composition Ethanol 10 - <25% , Methanol 1 - <2.5% , Glycerol 1 - <2.5%
Storage Requirements <20°C
Stain Type Gram Stain
Show More Show Less
compliance-icons
Product Identifier
  • Safranin
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P370+P378-In case of fire: Use to extinguish.
  • P403+P235-Store in a well-ventilated place. Keep cool.
  • P501-Dispose of contents/container to .
  • P240-Ground/bond container and receiving equipment.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P242-Use only non-sparking tools.
  • P243-Take precautionary measures against static discharge.
Supplemental information
  • MIXTURE LIST-Contains : ethanol, methanol