Learn More
Eppendorf SafeCode Plates
Product Code. 30130210 Shop All Eppendorf Products
Click to view available options
Color:
Blue
Green
Red
Unit Size:
Pack of 25
Product Code. 30130210
Brand: Eppendorf 0030113624
For optimized sample identification in the laboratory, the use of barcodes for the identification of plates is recommended. The SafeCode system for Eppendorf Plates is based on a pre-labeled 3-level coding for safe sample identification based on barcodes.
  • 3-level coding for safe sample and vessel identification
  • Codes on three sides of plates for flexible reading
  • ECC 200 error correction function enables safe code reading with up to 25 % damaged codes
  • Additional, ID-specific information on plates available via Eppendorf DataPort for documentation purposes
  • Direct import into eLabInventory and eLabJournal™ software for convenient handling
  • Import to other database systems possible
  • Manufactured from high-quality polypropylene for highest sample purity
TRUSTED_SUSTAINABILITY
Specifications
Barcode Alphanumeric
Format SBS Standard
Array 8 x 12
Packaging 25 Plates
Color Blue
Product Type PCR Plate
Skirt Style Skirted
Material Polypropylene, Polycarbonate
Volume (Metric) Well 150 μL
No. of Wells 96
Sterility Non-sterile
Surface Treatment Non-treated
Well Shape Round, Conical Bottom (V-well)
No. per Pack 25 Pk.
Well Color Colorless