missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ S1P3/EDG-3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30849-100ug
This item is not returnable.
View return policy
Description
S1P3/EDG-3 Polyclonal antibody specifically detects S1P3/EDG-3 in Human,Mouse samples. It is validated for Immunoprecipitation,Western Blot
Specifications
| S1P3/EDG-3 | |
| Polyclonal | |
| Western Blot : 5-10 μg/mL, Immunoprecipitation | |
| EDG-3, EDG3FLJ37523, Endothelial differentiation G-protein coupled receptor 3, endothelial differentiation, sphingolipid G-protein-coupled receptor, 3, FLJ93220, G protein-coupled receptor, endothelial differentiation gene-3, LPB3, MGC71696, S1P receptor 3, S1P receptor Edg-3, S1P receptor EDG3, S1P3, sphingosine 1-phosphate receptor 3, Sphingosine 1-phosphate receptor Edg-3, sphingosine-1-phosphate receptor 3 | |
| Rabbits were immunized with a synthetic peptide derived from the N terminal of the S1P3/EDG-3 protein. | |
| 100 μg | |
| Cell Biology, Cell Cycle and Replication, Cellular Markers, GPCR, Lipid and Metabolism, Signal Transduction | |
| 1903 | |
| Store at -20°C. Avoid freeze/thaw cycles. |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Phosphate buffered saline | |
| Rabbit | |
| Ammonium sulfate precipitation | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction